
Neodymium(III) trifluoromethanesulphonate ≥97.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:34622-08-7
:
Formula:C₃F₉NdO₉S₃
MDL Number:MFCD00192343
ADR:8,III
:591.45 g/mol
UN:1759
:GHS05
Specification Test Results
- Appearance:White to Amber to Dark purple powder to crystal
- Purity (Chelometric Titration):min. 97.0 % (calcd.on anh.substance)
- Water:max. 10.0 %
Specification Test Results
Frequently Bought Together