
Lanthanum(III) trifluoromethanesulphonate ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:52093-26-2
:
Formula:C₃F₉LaO₉S₃
MDL Number:MFCD00077560
:586.12 g/mol
UN:1759
ADR:8,III
:GHS05
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Chelometric Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together