Order Entry
Italy
ContactUsLinkComponent
2-Acetyl-7-methoxybenzofuran ≥98.0% (by GC)
2-Acetyl-7-methoxybenzofuran ≥98.0% (by GC)
Catalog # TCIAA1317-10G
Supplier:  TCI
CAS Number:  
undefined
2-Acetyl-7-methoxybenzofuran ≥98.0% (by GC)
Catalog # TCIAA1317-10G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 g
  • Seq:
    3236

Specifications

Specification Test Results

  • Appearance:
    White to Amber powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    92.0 to 95.0 deg-C
  • Solubility in hot Methanol:
    very faint turbidity

Specification Test Results

About this item

 :
 :190.2 g/mol
Boiling Pt:180°C
CAS Number:43071-52-9
Formula:C₁₁H₁₀O₃
MDL Number:MFCD00016911