Order Entry
Italy
ContactUsLinkComponent
Methyl-2,4-dichlorobenzoate ≥98.0% (by GC)
Methyl-2,4-dichlorobenzoate ≥98.0% (by GC)
  TCIAD2855-5G
 :  TCI
 :  
undefined
Methyl-2,4-dichlorobenzoate ≥98.0% (by GC)
  TCIAD2855-5G
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

 

 

  • Appearance:
    Light yellow to Yellow to Orange clear liquid
  • Purity (GC):
    min. 98.0 %

 

 

 :
 :205.04 g/mol
 :252-374-1
 :35112-28-8
 :C₈H₆Cl₂O₂
 :MFCD00085117