trans-1,2-Dichlorocyclohexane ≥96.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:822-86-6
:
Formula:C₆H₁₀Cl₂
MDL Number:MFCD00003824
:153.05 g/mol
Boiling Pt:69°C
Melting Pt:–6 °C
Density:1,1842 g/cm³ (20 °C)
Flash Pt:66°C
EINECS:212-503-4
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 96.0 %
Specification Test Results
Frequently Bought Together