
Cyclohexanecarboxylic acid ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:98-89-5
:
Formula:C₇H₁₂O₂
MDL Number:MFCD00001461
:128.17 g/mol
Merck Index:14,02724
Boiling Pt:232 °C (1013 hPa)
Melting Pt:31 °C
Density:1,03 g/cm³ (20 °C)
Flash Pt:110 °C
EINECS:202-711-3
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White or Colorless to Light yellow powder to lump to clear liquid
- Purity (GC):min. 96.0 %
- Purity (Neutralization titration):min. 98.0 %
Specification Test Results
Frequently Bought Together