
Cycloheptanone ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:502-42-1
:
Formula:C₇H₁₂O
MDL Number:MFCD00004159
:112.17 g/mol
Merck Index:14,02722
Boiling Pt:180°C
Melting Pt:–21°C
Density:0,949 g/cm³ (20 °C)
Flash Pt:56°C
Storage Temperature:Ambient
EINECS:207-937-6
UN:1224
ADR:3,III
:GHS02
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together