


Mesitylene ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:108-67-8
:
Formula:C₉H₁₂
MDL Number:MFCD00008538
Merck Index:14,05907
Boiling Pt:164°C
Melting Pt:–45 °C
Density:0,8637 g/cm³ (20 °C)
Flash Pt:50°C
Storage Temperature:Ambient
UN:2325
ADR:3,III
:120.19 g/mol
EINECS:203-604-4
:GHS02
:GHS07
:GHS09
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 97.0 %
Specification Test Results
Frequently Bought Together