



1,2,3-Trimethylbenzene ≥80.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:526-73-8
:
Formula:C₉H₁₂
MDL Number:MFCD00008520
:120.19 g/mol
Boiling Pt:176°C
Melting Pt:–25 °C
Density:0,894 g/cm³ (20 °C)
Flash Pt:44°C
EINECS:208-394-8
UN:3295
ADR:3,III
:GHS02
:GHS07
:GHS08
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 80.0 %
Specification Test Results
Frequently Bought Together