

p-Xylene ≥99.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:106-42-3
:
Formula:C₆H₄(CH₃)₂
MDL Number:MFCD00008556
:106.17 g/mol
Merck Index:14,10081
Boiling Pt:139°C
Melting Pt:13 °C
Density:1,096 g/cm³ (20 °C)
Flash Pt:27°C
Storage Temperature:Ambient
EINECS:203-396-5
UN:1307
ADR:3,III
:GHS02
:GHS07
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 99.0 %
Specification Test Results
Frequently Bought Together