Order Entry
Italy
ContactUsLinkComponent
2,5-Dimethylanisole ≥98.0% (by GC)
2,5-Dimethylanisole ≥98.0% (by GC)
Catalog # TCIAD2440-25G
Supplier:  TCI
CAS Number:  
undefined
2,5-Dimethylanisole ≥98.0% (by GC)
Catalog # TCIAD2440-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 98.0 %

Specification Test Results

About this item

 :
 :136.19 g/mol
Boiling Pt:194°C
Density:0,963 g/cm³ (20 °C)
Flash Pt:66°C
Storage Temperature:Ambient
EINECS:216-943-8
CAS Number:1706-11-2
Formula:C₉H₁₂O
MDL Number:MFCD00008378