Order Entry
Italy
ContactUsLinkComponent
 
2,6-Dimethylanisole ≥98.0% (by GC)
  TCIAD2383-25ML
 :  TCI
 :  
undefined
2,6-Dimethylanisole ≥98.0% (by GC)
  TCIAD2383-25ML
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    25 mL
  • Seq:
    2315

 

 

  • Appearance:
    Colorless to Light yellow to Light orange clear liquid
  • Purity (GC):
    min. 98.0 %

 

 

 :
 :136.19 g/mol
 :182 °C (1013 hPa)
 :0,96 g/cm³ (20 °C)
 :67°C
 :213-723-3
 :1004-66-6
 :C₉H₁₂O
 :MFCD00008380