Order Entry
Italy
ContactUsLinkComponent
2,3-Dimethylanisole ≥98.0% (by GC)
 
2,3-Dimethylanisole ≥98.0% (by GC)
  TCIAD2200-10ML
 :  TCI
 :  
undefined
 
2,3-Dimethylanisole ≥98.0% (by GC)
  TCIAD2200-10ML
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • Seq:
    2275

 

 

  • Appearance:
    White to Almost white powder to lump
  • Purity (GC):
    min. 98.0 %

 

 

 :
 :195 °C (1013 hPa)
 :29 °C
 :0,9596 g/cm³ (39.7 °C)
 :68°C
 :136.19 g/mol
 :220-948-0
 :GHS07
 :2944-49-2
 :C₉H₁₂O
 :MFCD00008376