
2,6-Diethylaniline ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:579-66-8
:
Formula:C₁₀H₁₅N
MDL Number:MFCD00007753
:149.24 g/mol
Boiling Pt:243 °C (1013 hPa)
Melting Pt:3.5 °C
Density:0,984 g/cm³ (20 °C)
Flash Pt:115°C
EINECS:209-445-7
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Colorless to Red to Green clear liquid
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together