
2,4-Dimethylbenzoic acid ≥99.0% (by titrimetric analysis)
Catalog # TCIAD1438-10G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 g
- Seq:3236
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):min. 99.0 %
- Melting point:124.0 to 127.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
About this item
:
:150.18 g/mol
Boiling Pt:267 °C (1013 hPa)
Melting Pt:123…124 °C
EINECS:210-246-2
:GHS07
Storage Temperature:Ambient
CAS Number:611-01-8
Formula:C₉H₁₀O₂
MDL Number:MFCD00002480