Order Entry
Italy
ContactUsLinkComponent
1,3-Diphenylurea ≥98.0% (by HPLC, total nitrogen)
1,3-Diphenylurea ≥98.0% (by HPLC, total nitrogen)
Supplier:  TCI
Certificates
undefined
1,3-Diphenylurea ≥98.0% (by HPLC, total nitrogen)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (with Total Nitrogen):
    min. 98.0 %
  • Solubility in hot Acetic acid:
    almost transparency

Specification Test Results

Related Information

Frequently Bought Together