


Isopentyl nitrite (mixed isomers ) ≥95.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:110-46-3
:
Formula:C₅H₁₁NO₂
MDL Number:MFCD00002057
:117.15 g/mol
Merck Index:14,05123
Boiling Pt:99°C
Flash Pt:3°C
EINECS:203-770-8
UN:1113
ADR:3,II
:GHS02
:GHS06
:GHS08
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Light yellow to Yellow clear liquid
- Purity (GC):min. 95.0 %
Specification Test Results
Frequently Bought Together