6-Methoxy-2-naphthaldehyde ≥97.0% (by GC)
: TCI
:3453-33-6
:
:C₁₂H₁₀O₂
:MFCD00081152
:196°C
:186.21 g/mol
:Refrigerator
:222-377-2
- Environmentally Preferable:
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (GC):min. 97.0 %
- Melting point:81.0 to 84.0 deg-C
Frequently Bought Together