
3,3-Diphenylpropylamine ≥98.0% (by titrimetric analysis)
Catalog # TCIAD1855-25G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Orange to Green powder to lump
- Purity (Nonaqueous Titration):min. 98.0 %
- Freezing point:24.0 to 29.0 deg-C
Specification Test Results
About this item
:
:211.31 g/mol
EINECS:226-984-3
:GHS07
CAS Number:5586-73-2
Formula:C₁₅H₁₇N
MDL Number:MFCD00008202