Order Entry
Italy
Orders LinkContactUsLinkComponent
2-Butanone oxime ≥99.0% (by GC)

2-Butanone oxime ≥99.0% (by GC)

Corrosive
Irritant
Health
Supplier:   TCI
2-Butanone oxime ≥99.0% (by GC)
Supplier:  TCI
Certificates
2-Butanone oxime ≥99.0% (by GC)

2 Options Available Below

Product Details & Documents
  • CAS Number:96-29-7
  • Formula:C₄H₉NO
  • MDL Number:MFCD00013935
  • Molecular Weight:87.12 g/mol
  • Boiling Pt:153°C
  • Melting Pt:–30°C
  • Melting Pt:-30°C
  • Flash Pt:62°C
  • EINECS:202-496-6
  • UN:2810
  • ADR:6.1,II
Specifications for Product Family

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 99.0 %
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    25 mL
    Log in for availability
    Log in for availability
    1 * 25 mL
    1 * 25 mL - 16,00 €
    16,00 €
    1 * 25 mL
       16,00 €

  • Glass bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 24,00 €
    24,00 €
    1 * 500 mL
       24,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.