


2-sec-Butylphenol ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:89-72-5
:
Formula:C₁₀H₁₄O
MDL Number:MFCD00002225
Boiling Pt:228°C
Melting Pt:12.2 °C
Density:0,9981 g/cm³ (20 °C)
Flash Pt:102°C
Storage Temperature:Ambient
EINECS:201-933-8
ADR:8,III
:150.22 g/mol
UN:3145
:GHS05
:GHS07
:GHS09
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 97.0 %
Specification Test Results
Frequently Bought Together