Order Entry
Italy
ContactUsLinkComponent
 
 
 
2,6-Dinitrotoluene ≥99.0% (by GC)
  TCIAD1151-25G
 :  TCI
 :  
undefined
 
 
2,6-Dinitrotoluene ≥99.0% (by GC)
  TCIAD1151-25G
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

 

 

  • Appearance:
    White to Orange to Green powder to crystal
  • Purity (GC):
    min. 99.0 %
  • Melting point:
    57.0 to 65.0 deg-C
  • Solubility in Methanol:
    almost transparency

 

 

 :
 :182.14 g/mol
 :300 °C (1013 hPa)
 :65.2 °C
 :1,538 g/cm³ (15 °C)
 :Ambient
 :210-106-0
 :3454
 :6.1,II
 :GHS06
 :GHS08
 :606-20-2
 :C₇H₆N₂O₄
 :MFCD00007158