
2-Fluoro-5-nitrobenzotrifluoride ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:400-74-8
:
Formula:C₇H₃F₄NO₂
MDL Number:MFCD00024576
:209.1 g/mol
Boiling Pt:110°C
Melting Pt:22…24 °C
Density:1,52 g/cm³ (20 °C)
Flash Pt:91°C
EINECS:206-925-8
ADR:6.1,III
:GHS07
Specification Test Results
- Appearance:Light yellow to Amber to Dark green clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together