

α,α,α,4-Tetrafluorotoluene ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:402-44-8
:
Formula:C₇H₄F₄
MDL Number:MFCD00000399
:164.1 g/mol
Boiling Pt:103°C
Density:1,293 g/cm³ (25 °C)
Flash Pt:11°C
Storage Temperature:Refrigerator
EINECS:206-944-1
UN:1993
ADR:3,II
:GHS02
:GHS07
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together