Order Entry
Italy
ContactUsLinkComponent
 
 
 
5-Chloro-2-nitrobenzotrifluoride ≥80.0% (by GC)
  TCIAC0226-25G
 :  TCI
 :  
undefined
 
 
5-Chloro-2-nitrobenzotrifluoride ≥80.0% (by GC)
  TCIAC0226-25G
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

 

 

  • Appearance:
    Light yellow to Yellow to Green clear liquid
  • Purity (GC):
    min. 80.0 %

 

 

 :
 :225.55 g/mol
 :224°C
 :21…22 °C
 :1,526 g/cm³ (20 °C)
 :102°C
 :204-280-7
 :2810
 :6.1,III
 :GHS06
 :GHS09
 :118-83-2
 :C₇H₃ClF₃NO₂
 :MFCD00007298