
2-Chloro-5-nitrobenzotrifluoride ≥97.0% (by GC)
Catalog # TCIAC0923-25G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White or Colorless to Yellow powder to lump to clear liquid
- Purity (GC):min. 97.0 %
- Freezing point:20.0 to 23.0 deg-C
Specification Test Results
About this item
:
:225.55 g/mol
Boiling Pt:232 °C (1013 hPa)
Melting Pt:25…27 °C
Density:1,527 g/cm³ (20 °C)
EINECS:212-287-1
UN:2811
ADR:6.1,III
:GHS06
CAS Number:777-37-7
Formula:C₇H₃ClF₃NO₂
MDL Number:MFCD00007296