Order Entry
Italy
ContactUsLinkComponent
4-Chloro-3-nitrobenzotrifluoride ≥97.0% (by GC)
Irritant
4-Chloro-3-nitrobenzotrifluoride ≥97.0% (by GC)
Supplier:  TCI
Certificates
undefined
Irritant
4-Chloro-3-nitrobenzotrifluoride ≥97.0% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Light orange to Yellow to Green clear liquid
  • Purity (GC):
    min. 97.0 %

Specification Test Results

Frequently Bought Together