

4-Chloro-α,α,α-trifluorotoluene ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:98-56-6
:
Formula:C₇H₄ClF₃
MDL Number:MFCD00000627
:180.56 g/mol
Merck Index:14,02126
Boiling Pt:140°C
Melting Pt:–36 °C
Density:1,346 g/cm³ (20 °C)
Flash Pt:43°C
Storage Temperature:Refrigerator
EINECS:202-681-1
UN:2234
ADR:3,III
:GHS02
:GHS07
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Related Information
Frequently Bought Together