


α,2-Dichlorotoluene ≥99.0% (by GC)
: TCI
:611-19-8
:
:C₇H₆Cl₂
:MFCD00000893
:161.03 g/mol
:97°C
:–13 °C
:1,2902 g/cm³ (20 °C)
:117°C
:Refrigerator
:210-258-8
:2235
:6.1,III
:GHS07
:GHS08
:GHS09
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 99.0 %
Frequently Bought Together