Order Entry
Italy
ContactUsLinkComponent
α,3,4-Trichlorotoluene ≥98.0% (by GC)
Corrosive
Irritant
α,3,4-Trichlorotoluene ≥98.0% (by GC)
Catalog # TCIAT0401-25G
Supplier:  TCI
CAS Number:  
undefined
Corrosive
Irritant
α,3,4-Trichlorotoluene ≥98.0% (by GC)
Catalog # TCIAT0401-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 98.0 %

Specification Test Results

About this item

 :
 :195.47 g/mol
Boiling Pt:241°C
Melting Pt:37.5 °C
Density:1,41 g/cm³ (20 °C)
Flash Pt:155°C
Storage Temperature:Ambient
EINECS:203-033-0
UN:3265
ADR:8,III
 :GHS05
 :GHS07
CAS Number:102-47-6
Formula:C₇H₅Cl₃
MDL Number:MFCD00000906