2,5-Dichlorotoluene ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:19398-61-9
:
Formula:C₇H₆Cl₂
MDL Number:MFCD00000609
:161.03 g/mol
Boiling Pt:199°C
Density:1,2535 g/cm³ (20 °C)
Flash Pt:111°C
EINECS:243-032-2
ADR:9,III
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together