
3,4-Dichlorotoluene ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:95-75-0
:
Formula:C₇H₆Cl₂
MDL Number:MFCD00000556
:161.03 g/mol
Boiling Pt:205°C
Melting Pt:–15.2 °C
Density:1,2564 g/cm³ (20 °C)
Flash Pt:118°C
Storage Temperature:Ambient
EINECS:202-447-9
UN:3082
ADR:9,III
:GHS09
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together