
Isovalerophenone ≥99.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:582-62-7
:
Formula:(C₆H₅)C(O)CH₂CH(CH₃)CH₃
MDL Number:MFCD00026486
:162.23 g/mol
Boiling Pt:236°C
Melting Pt:2 °C
Density:0,966 g/cm³ (25 °C)
Flash Pt:99°C
EINECS:209-489-7
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 99.0 %
Specification Test Results
Frequently Bought Together