4-tert-Butylbenzaldehyde
Supplier: Thermo Scientific
Certificates
About this item
Synonyms: 4-(tert-Butyl)benzaldehyde
CAS Number:939-97-9
:
Formula:C₁₁H₁₄O
MDL Number:MFCD00035742
Boiling Pt:58…60 °C (0.2 mmHg)
Density:0.971 g/cm³ (20 °C)
Molecular Weight:162.23 g/mol
Storage Temperature:Ambient
EINECS:213-367-9
UN:3082
ADR:9 III
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together