

4-Bromo-α,α,α-trifluorotoluene ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:402-43-7
:
Formula:C₇H₄BrF₃
MDL Number:MFCD00000398
Boiling Pt:155°C
Density:1,6347 g/cm³ (20 °C)
Flash Pt:48°C
EINECS:206-943-6
UN:1993
ADR:3,III
:225.01 g/mol
:GHS02
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Related Information
Frequently Bought Together