Order Entry
Italy
ContactUsLinkComponent
 
 
3-Bromo-4-chlorobenzotrifluoride ≥97.0% (by GC)
  TCIAB2743-25G
 :  TCI
 :  
undefined
 
3-Bromo-4-chlorobenzotrifluoride ≥97.0% (by GC)
  TCIAB2743-25G
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

 

 

  • Appearance:
    Colorless to Light yellow clear liquid
  • Purity (GC):
    min. 97.0 %

 

 

 :
 :259.45 g/mol
 :190°C
 :-22°C
 :–22°C
 :1,743 g/cm³ (25 °C)
 :94°C
 :207-226-0
 :GHS07
 :454-78-4
 :C₇H₃BrClF₃
 :MFCD00018093