
α,4-Dibromotoluene ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:589-15-1
:
Formula:C₇H₆Br₂
MDL Number:MFCD00000179
:249.93 g/mol
Merck Index:14,01409
Boiling Pt:129 - 130 °C (18 -rr)
Melting Pt:60 °C
Storage Temperature:Ambient
EINECS:209-636-5
UN:3261
ADR:8,III
:GHS05
Specification Test Results
- Appearance:White to Almost white powder to crystaline
- Purity (GC):min. 98.0 %
- Melting point:60.0 to 64.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together