(S)-(-)-α-Methoxy-α-(trifluoromethyl)phenylacetic acid ≥98.0% (by GC, titration analysis) optical resolving
Supplier: TCI
Certificates
About this item
CAS Number:17257-71-5
:
Formula:C₁₀H₉F₃O₃
MDL Number:MFCD00064200
:234.17 g/mol
EINECS:241-292-1
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to lump
- Purity (GC):min. 98.0 %
- Optical purity (GC):min. 99.0 ee%
- Purity (Neutralization titration):min. 98.0 %
- Specific rotation [a]20/D:-70.0 to -75.0 deg (C=2, Methanol)
Specification Test Results
Frequently Bought Together