Order Entry
Italy
ContactUsLinkComponent
1,2,4-Trimethoxybenzene ≥97.0% (by GC)
1,2,4-Trimethoxybenzene ≥97.0% (by GC)
Catalog # TCIAT1130-25G
Supplier:  TCI
CAS Number:  
undefined
1,2,4-Trimethoxybenzene ≥97.0% (by GC)
Catalog # TCIAT1130-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    Light yellow to Brown clear liquid
  • Purity (GC):
    min. 97.0 %

Specification Test Results

About this item

 :
Merck Index:13,01073
Boiling Pt:247°C
Melting Pt:19…21 °C
Density:1,126 g/cm³ (20 °C)
 :168.19 g/mol
Storage Temperature:Ambient
EINECS:205-219-7
CAS Number:135-77-3
Formula:C₉H₁₂O₃
MDL Number:MFCD00008360