1,3-Diethoxybenzene ≥97.0% (by GC)
TCIAD0453-25G
: TCI
:
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 97.0 %
:
:166.22 g/mol
:235°C
:218-071-3
:2049-73-2
:C₁₀H₁₄O₂
:MFCD00015145