
2-(4-Chloro-3-nitrobenzoyl)benzoic acid ≥97.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:85-54-1
:
Formula:C₁₄H₈ClNO₅
MDL Number:MFCD00007082
:305.67 g/mol
EINECS:201-613-8
:GHS07
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 97.0 area%
- Purity (Neutralization titration):min. 97.0 %
- Melting point:198.0 to 202.0 deg-C
Specification Test Results
Frequently Bought Together