
1-Butyl-4-methylpyridinium bromide ≥98.0% (by HPLC, titration analysis)
: TCI
:65350-59-6
:
:C₁₀H₁₆BrN
:MFCD06798209
:230.15 g/mol
:GHS07
- Appearance:White to Yellow to Orange powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:135.0 to 139.0 deg-C
Frequently Bought Together