Order Entry
Italy
ContactUsLinkComponent
1-Ethyl-3-methylimidazolium methanesulphonate ≥98.0% (by total nitrogen basis)
Corrosive
1-Ethyl-3-methylimidazolium methanesulphonate ≥98.0% (by total nitrogen basis)
Supplier:  TCI
Certificates
undefined
Corrosive
1-Ethyl-3-methylimidazolium methanesulphonate ≥98.0% (by total nitrogen basis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Light yellow to Brown clear liquid
  • Purity (HPLC):
    min. 97.0 area%
  • Purity (with Total Nitrogen):
    min. 98.0 %

Specification Test Results

Related Information

Frequently Bought Together