

5-Chlorooxindole ≥97.0% (by GC, total nitrogen)
Supplier: TCI
Certificates
About this item
CAS Number:17630-75-0
:
Formula:C₈H₆ClNO
MDL Number:MFCD00191927
:167.59 g/mol
EINECS:412-200-9
:GHS07
:GHS08
Specification Test Results
- Appearance:Light yellow to Brown powder to crystal
- Purity (GC):min. 97.0 %
- Purity (with Total Nitrogen):min. 97.0 %
- Melting point:194.0 to 199.0 deg-C
- Solubility in hot Methanol:almost transparency
Specification Test Results
Frequently Bought Together