
Ethyl piperazine-1-carboxylate ≥96.0% (by GC)
Catalog # TCIAP1349-25G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:Light orange to Yellow to Green clear liquid
- Purity (GC):min. 96.0 %
Specification Test Results
About this item
:
:158.2 g/mol
Boiling Pt:237°C
EINECS:204-395-2
:GHS07
CAS Number:120-43-4
Formula:C₇H₁₄N₂O₂
MDL Number:MFCD00005964