Erythrosin B ≥95.0% (by HPLC)
Catalog # TCIAT0124-25G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:Light yellow to Brown powder to crystal
- Purity (HPLC):min. 95.0 area%
- Solubility in EtOH (95vol%):almost transparency
- Sensitiveness:to pass test
Specification Test Results
About this item
:
:835.89 g/mol
EINECS:240-046-0
CAS Number:15905-32-5
Formula:C₂₀H₈I₄O₅
MDL Number:MFCD00005044