
1-Methyl-3-octylimidazolium chloride ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:64697-40-1
:
Formula:C₁₂H₂₃ClN₂
MDL Number:MFCD03095432
:230.78 g/mol
Melting Pt:12°C
:GHS07
Specification Test Results
- Appearance:Colorless to Yellow to Orange clear liquid
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together