


Mesityl oxide ≥95.0% (by GC)
: TCI
:141-79-7
:
:C₆H₁₀O
:MFCD00008900
:14,05908
:130°C
:27°C
:Ambient
:205-502-5
:1229
:3,III
:98.14 g/mol
:GHS02
:GHS06
:GHS08
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 95.0 %
Frequently Bought Together