


1,4-Dinitrobenzene ≥99.0% (by GC)
: TCI
:100-25-4
:
:C₆H₄N₂O₄
:MFCD00007314
:168.11 g/mol
:14,03273
:299 °C (1013 hPa)
:172 °C
:1,625 g/cm³ (20 °C)
:Ambient
:202-833-7
:3443
:6.1,II
:GHS06
:GHS08
:GHS09
- Appearance:Light yellow to Brown powder to crystaline
- Purity (GC):min. 99.0 %
- Melting point:171.0 to 175.0 deg-C
- Solubility in Acetonitrile:very faint turbidity
Frequently Bought Together