
3,4,5-Trimethoxyaniline ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:24313-88-0
:
Formula:C₉H₁₃NO₃
MDL Number:MFCD00008393
:183.21 g/mol
Boiling Pt:293 °C (1013 hPa)
Melting Pt:110…113 °C
Storage Temperature:Ambient
EINECS:246-154-4
ADR:6.1,III
:GHS07
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:111.0 to 115.0 deg-C
Specification Test Results
Frequently Bought Together